N-(3,4-Dimethylphenyl)-4-hydrazino-4-oxobutanamide

Supplier: Matrix Scientific

023980-500MG
101817-162EA 306.15 USD
101817-162
N-(3,4-Dimethylphenyl)-4-hydrazino-4-oxobutanamide
N-(3,4-Dimethylphenyl)-4-hydrazino-4-oxobutanamide

Hazard: Irritant

Formula: C₁₂H₁₇N₃O₂
MDL Number: MFCD02603991
CAS Number: 328025-45-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR