N-(2,4-Dichlorophenyl)-4-hydrazino-4-oxobutanamide

Supplier: Matrix Scientific

023979-500MG
101817-160EA 228.85 USD
101817-160
N-(2,4-Dichlorophenyl)-4-hydrazino-4-oxobutanamide
N-(2,4-Dichlorophenyl)-4-hydrazino-4-oxobutanamide

Hazard: Irritant

Formula: C₁₀H₁₁Cl₂N₃O₂
MDL Number: MFCD02188787
CAS Number: 328025-23-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR