N-(4-Ethoxyphenyl)-4-hydrazino-4-oxobutanamide

Supplier: Matrix Scientific

023976-500MG
101817-154EA 253.85 USD
101817-154
N-(4-Ethoxyphenyl)-4-hydrazino-4-oxobutanamide
N-(4-Ethoxyphenyl)-4-hydrazino-4-oxobutanamide

Hazard: Irritant

Formula: C₁₂H₁₇N₃O₃
MDL Number: MFCD02188621
CAS Number: 315672-07-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR