N-[4-(Hydrazinocarbonyl)phenyl]-2-methylbenzamide

Supplier: Matrix Scientific

023975-500MG
101817-152EA 253.85 USD
101817-152
N-[4-(Hydrazinocarbonyl)phenyl]-2-methylbenzamide
N-[4-(Hydrazinocarbonyl)phenyl]-2-methylbenzamide

Hazard: Irritant

Formula: C₁₅H₁₅N₃O₂
MDL Number: MFCD02188619
CAS Number: 315671-80-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR