4-(2,4-Dichlorophenoxy)butanohydrazide
Supplier: Matrix Scientific
Synonyms:
4-(2,4-Dichlorophenoxy)butanoic acid hydrazide
023972-500MG
101817-146EA
135.38
USD
101817-146
4-(2,4-Dichlorophenoxy)butanohydrazide
4-(2,4-Dichlorophenoxy)butanohydrazide
Hazard: Irritant
Formula:
C₁₀H₁₂Cl₂N₂O₂ MW: 263.13 g/mol |
MDL Number:
MFCD01248815 CAS Number: 131426-24-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...