2-(2,4-Dichlorophenoxy)propionic acid hydrazide

Supplier: Matrix Scientific
Warning

023971-500MG
101817-144EA 11.5 USD
101817-144
2-(2,4-Dichlorophenoxy)propionic acid hydrazide
2-(2,4-Dichlorophenoxy)propionic acid hydrazide
Formula: C₉H₁₀Cl₂N₂O₂
MDL Number: MFCD00204152
CAS Number: 15253-89-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR