N-Benzyl-4-hydrazino-4-oxobutanamide

Supplier: Matrix Scientific

023968-500MG
101817-138EA 306.15 USD
101817-138
N-Benzyl-4-hydrazino-4-oxobutanamide
N-Benzyl-4-hydrazino-4-oxobutanamide

Hazard: Irritant

Formula: C₁₁H₁₅N₃O₂
MDL Number: MFCD02105445
CAS Number: 315249-23-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR