1-Amino-4,6-dimethyl-2-oxo-1,2-dihydro-3-pyridinecarbonitrile

Supplier: Matrix Scientific

023966-500MG
101817-134EA 97.75 USD
101817-134
1-Amino-4,6-dimethyl-2-oxo-1,2-dihydro-3-pyridinecarbonitrile
1-Amino-4,6-dimethyl-2-oxo-1,2-dihydro-3-pyridinecarbonitrile

Hazard: Irritant

Formula: C₈H₉N₃O
Melting Pt: 174 °C
MDL Number: MFCD00086187
CAS Number: 1562-12-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR