2-[(1-Bromo-2-naphthyl)oxy]acetohydrazide

Supplier: Matrix Scientific

023965-500MG
101817-132EA 112.7 USD
101817-132
2-[(1-Bromo-2-naphthyl)oxy]acetohydrazide
2-[(1-Bromo-2-naphthyl)oxy]acetohydrazide

Hazard: Irritant

Formula: C₁₂H₁₁BrN₂O₂
MDL Number: MFCD00245428
CAS Number: 315248-38-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR