2-[1-(1-Naphthylmethyl)-3-oxo-2-piperazinyl]acetic acid
Supplier: Matrix Scientific
023963-500MG
101817-128EA
306.15
USD
101817-128
2-[1-(1-Naphthylmethyl)-3-oxo-2-piperazinyl]acetic acid
2-[1-(1-Naphthylmethyl)-3-oxo-2-piperazinyl]acetic acid
Hazard: Irritant
Formula:
C₁₇H₁₈N₂O₃ |
MDL Number:
MFCD05666811 CAS Number: 1093946-38-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...