2-(2-Chloroanilino)acetohydrazide

Supplier: Matrix Scientific

023959-500MG
101817-120EA 306.15 USD
101817-120
2-(2-Chloroanilino)acetohydrazide
2-(2-Chloroanilino)acetohydrazide

Hazard: Irritant

Formula: C₈H₁₀ClN₃O
MDL Number: MFCD02006273
CAS Number: 2371-29-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR