2-(4-Methoxyanilino)acetohydrazide
Supplier: Matrix Scientific
023956-500MG
101817-114EA
109.23
USD
101817-114
2-(4-Methoxyanilino)acetohydrazide
2-(4-Methoxyanilino)acetohydrazide
Hazard: Irritant
Formula:
C₉H₁₃N₃O₂ |
MDL Number:
MFCD00462929 CAS Number: 79476-73-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...