2-(4-Ethoxyanilino)acetohydrazide

Supplier: Matrix Scientific

023951-500MG
101817-104EA 189.75 USD
101817-104
2-(4-Ethoxyanilino)acetohydrazide
2-(4-Ethoxyanilino)acetohydrazide

Hazard: Irritant

Formula: C₁₀H₁₅N₃O₂
MDL Number: MFCD02255627
CAS Number: 100133-39-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR