2,2-Diphenylacetohydrazide

Supplier: Matrix Scientific

023948-500MG
101817-098EA 140.3 USD
101817-098
2,2-Diphenylacetohydrazide
2,2-Diphenylacetohydrazide

Hazard: Irritant

Formula: C₁₄H₁₄N₂O
Melting Pt: 135 °C
MDL Number: MFCD00544954
CAS Number: 6636-02-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR