N-[4-(Hydrazinocarbonyl)phenyl]propanamide

Supplier: Matrix Scientific

023945-500MG
101817-092EA 306.15 USD
101817-092
N-[4-(Hydrazinocarbonyl)phenyl]propanamide
N-[4-(Hydrazinocarbonyl)phenyl]propanamide

Hazard: Irritant

Formula: C₁₀H₁₃N₃O₂
MDL Number: MFCD01814010
CAS Number: 349642-97-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR