2-(2-Isopropylphenoxy)acetohydrazide
Supplier: Matrix Scientific
023937-500MG
101817-076EA
306.15
USD
101817-076
2-(2-Isopropylphenoxy)acetohydrazide
2-(2-Isopropylphenoxy)acetohydrazide
Hazard: Irritant
Formula:
C₁₁H₁₆N₂O₂ |
MDL Number:
MFCD03423119 CAS Number: 501111-57-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...