2-([1,1'-Biphenyl]-2-yloxy)acetohydrazide

Supplier: Matrix Scientific

023936-500MG
101817-074EA 228.85 USD
101817-074
2-([1,1'-Biphenyl]-2-yloxy)acetohydrazide
2-([1,1'-Biphenyl]-2-yloxy)acetohydrazide

Hazard: Irritant

Formula: C₁₄H₁₄N₂O₂
MDL Number: MFCD00980733
CAS Number: 5493-64-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR