2-(4-Chloro-3,5-dimethylphenoxy)acetohydrazide

Supplier: Matrix Scientific

023935-500MG
101817-072EA 140.3 USD
101817-072
2-(4-Chloro-3,5-dimethylphenoxy)acetohydrazide
2-(4-Chloro-3,5-dimethylphenoxy)acetohydrazide

Hazard: Irritant

Formula: C₁₀H₁₃ClN₂O₂
MDL Number: MFCD00705188
CAS Number: 156867-62-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR