2-(4-Chloro-3-methylphenoxy)acetohydrazide

Supplier: Matrix Scientific

023934-500MG
101817-070EA 53.85 USD
101817-070
2-(4-Chloro-3-methylphenoxy)acetohydrazide
2-(4-Chloro-3-methylphenoxy)acetohydrazide

Hazard: Irritant

Formula: C₉H₁₁ClN₂O₂
MDL Number: MFCD00705189
CAS Number: 72293-68-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR