Isopentyl-2-(3-oxo-2-piperazinyl)acetate

Supplier: Matrix Scientific

023932-500MG
101817-066EA 306.15 USD
101817-066
Isopentyl-2-(3-oxo-2-piperazinyl)acetate
Isopentyl-2-(3-oxo-2-piperazinyl)acetate

Hazard: Irritant

Formula: C₁₁H₂₀N₂O₃
MDL Number: MFCD08700370
CAS Number: 1032338-42-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR