(2-Chlorophenoxy)acetic acid
Supplier: Matrix Scientific
Synonyms:
2-CPA
023930-500MG
101817-062EA
15.38
USD
101817-062
(2-Chlorophenoxy)acetic acid
(2-Chlorophenoxy)acetic acid
Hazard: Irritant
Formula:
(C₆H₄Cl)OCH₂COOH MW: 186.59 g/mol |
MDL Number:
MFCD00004298 CAS Number: 614-61-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...