3-(1-Naphthyl)acrylic acid

Supplier: Matrix Scientific

Synonyms: 3-(Naphthalen-1-yl)acrylic acid

023929-1G
101817-060EA 218.46 USD
101817-060
3-(1-Naphthyl)acrylic acid
3-(1-Naphthyl)acrylic acid

Hazard: Irritant

Formula: C₁₃H₁₀O₂
MW: 198.22 g/mol
MDL Number: MFCD00014317
CAS Number: 13026-12-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR