Butyl-2-(3-oxo-2-piperazinyl)acetate

Supplier: Matrix Scientific

023926-500MG
101817-054EA 228.85 USD
101817-054
Butyl-2-(3-oxo-2-piperazinyl)acetate
Butyl-2-(3-oxo-2-piperazinyl)acetate

Hazard: Irritant

Formula: C₁₀H₁₈N₂O₃
MDL Number: MFCD02942036
CAS Number: 90973-88-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR