2-Amino-N-ethylbenzamide

Supplier: Matrix Scientific

023923-500MG
101817-048EA 228.85 USD
101817-048
2-Amino-N-ethylbenzamide
2-Amino-N-ethylbenzamide

Hazard: Irritant

Formula: C₉H₁₂N₂O
MDL Number: MFCD00957005
CAS Number: 32212-29-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR