Cyclohexyl-2-(3-oxo-2-piperazinyl)acetate

Supplier: Matrix Scientific

023919-500MG
101817-040EA 180 USD
101817-040
Cyclohexyl-2-(3-oxo-2-piperazinyl)acetate
Cyclohexyl-2-(3-oxo-2-piperazinyl)acetate

Hazard: Irritant

Formula: C₁₂H₂₀N₂O₃
MDL Number: MFCD05870566
CAS Number: 1008015-40-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR