2-Amino-N-isopropylbenzamide
Supplier: Matrix Scientific
023915-500MG
101817-030EA
78.46
USD
101817-030
2-Amino-N-isopropylbenzamide
2-Amino-N-isopropylbenzamide
Hazard: Irritant
Formula:
C₁₀H₁₄N₂O |
MDL Number:
MFCD00143620 CAS Number: 30391-89-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...