2-Amino-N-(3,4-dimethylphenyl)benzamide

Supplier: Matrix Scientific

023914-500MG
101817-028EA 180 USD
101817-028
2-Amino-N-(3,4-dimethylphenyl)benzamide
2-Amino-N-(3,4-dimethylphenyl)benzamide

Hazard: Irritant

Formula: C₁₅H₁₆N₂O
MDL Number: MFCD00121517
CAS Number: 102630-81-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR