(2-Aminophenyl)(4-methyl-1-piperazinyl)methanone

Supplier: Matrix Scientific

Synonyms: (2-Aminophenyl)(4-methylpiperazin-1-yl)methanone

023913-500MG
101817-026EA 90.77 USD
101817-026
(2-Aminophenyl)(4-methyl-1-piperazinyl)methanone
(2-Aminophenyl)(4-methyl-1-piperazinyl)methanone

Hazard: Irritant

Formula: C₁₂H₁₇N₃O
MW: 219.29 g/mol
Melting Pt: 76…79 °C
MDL Number: MFCD03013018
CAS Number: 93288-86-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR