2-Amino-N-(4-ethoxyphenyl)benzamide

Supplier: Matrix Scientific

023910-500MG
101817-018EA 167.69 USD
101817-018
2-Amino-N-(4-ethoxyphenyl)benzamide
2-Amino-N-(4-ethoxyphenyl)benzamide

Hazard: Irritant

Formula: C₁₅H₁₆N₂O₂
MDL Number: MFCD00578809
CAS Number: 19562-43-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR