2-Amino-N-cyclohexyl-N-methylbenzamide

Supplier: Matrix Scientific

023906-500MG
101817-010EA 228.85 USD
101817-010
2-Amino-N-cyclohexyl-N-methylbenzamide
2-Amino-N-cyclohexyl-N-methylbenzamide

Hazard: Irritant

Formula: C₁₄H₂₀N₂O
MDL Number: MFCD01929085
CAS Number: 73393-40-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR