(2-Aminophenyl)(1-azepanyl)methanone

Supplier: Matrix Scientific

023905-500MG
101817-008EA 134.55 USD
101817-008
(2-Aminophenyl)(1-azepanyl)methanone
(2-Aminophenyl)(1-azepanyl)methanone

Hazard: Irritant

Formula: C₁₃H₁₈N₂O
MDL Number: MFCD01238548
CAS Number: 159180-54-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR