2-Amino-N-(1-phenylethyl)benzamide

Supplier: Matrix Scientific

023902-500MG
101817-002EA 55.38 USD
101817-002
2-Amino-N-(1-phenylethyl)benzamide
2-Amino-N-(1-phenylethyl)benzamide

Hazard: Irritant

Formula: C₁₅H₁₆N₂O
MDL Number: MFCD00115985
CAS Number: 85592-80-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR