4-Amino-N-(2,6-dimethylphenyl)benzenesulfonamide

Supplier: Matrix Scientific

023901-500MG
101817-000EA 180 USD
101817-000
4-Amino-N-(2,6-dimethylphenyl)benzenesulfonamide
4-Amino-N-(2,6-dimethylphenyl)benzenesulfonamide

Hazard: Irritant

Formula: C₁₄H₁₆N₂O₂S
MDL Number: MFCD00124573
CAS Number: 6973-47-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR