4-Amino-N-propylbenzenesulfonamide

Supplier: Matrix Scientific

023898-500MG
101816-994EA 38.46 USD
101816-994
4-Amino-N-propylbenzenesulfonamide
4-Amino-N-propylbenzenesulphonamide

Hazard: Irritant

Formula: C₉H₁₄N₂O₂S
MW: 214.2900 g/mol
MDL Number: MFCD05666729
CAS Number: 58687-83-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR