N-Allyl-2-aminobenzamide
Supplier: Matrix Scientific
023897-500MG
101816-992EA
203.08
USD
101816-992
N-Allyl-2-aminobenzamide
N-Allyl-2-aminobenzamide
Hazard: Irritant
Formula:
C₁₀H₁₂N₂O |
MDL Number:
MFCD00456224 CAS Number: 4943-82-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...