N-Benzyl-2-(methylamino)benzamide

Supplier: Matrix Scientific

023894-500MG
101816-986EA 134.55 USD
101816-986
N-Benzyl-2-(methylamino)benzamide
N-Benzyl-2-(methylamino)benzamide

Hazard: Irritant

Formula: C₁₅H₁₆N₂O
MDL Number: MFCD01196807
CAS Number: 56042-78-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR