4-Amino-N,N-dipropylbenzenesulfonamide

Supplier: Matrix Scientific

023887-500MG
101816-972EA 228.85 USD
101816-972
4-Amino-N,N-dipropylbenzenesulfonamide
4-Amino-N,N-dipropylbenzenesulfonamide

Hazard: Irritant

Formula: C₁₂H₂₀N₂O₂S
MDL Number: MFCD05666700
CAS Number: 21646-92-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR