Benzyl-2-(3-oxo-2-piperazinyl)acetate

Supplier: Matrix Scientific

023884-500MG
101816-966EA 306.15 USD
101816-966
Benzyl-2-(3-oxo-2-piperazinyl)acetate
Benzyl-2-(3-oxo-2-piperazinyl)acetate

Hazard: Irritant

Formula: C₁₃H₁₆N₂O₃
MW: 248.28 g/mol
MDL Number: MFCD05666693
CAS Number: 1093760-28-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR