Methyl-2-(3-oxo-2-piperazinyl)acetate

Supplier: Matrix Scientific

023883-500MG
101816-964EA 306.15 USD
101816-964
Methyl-2-(3-oxo-2-piperazinyl)acetate
Methyl-2-(3-oxo-2-piperazinyl)acetate

Hazard: Irritant

Formula: C₇H₁₂N₂O₃
MDL Number: MFCD03011617
CAS Number: 89852-17-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR