4-Amino-N-methyl-N-phenylbenzenesulfonamide

Supplier: Matrix Scientific

023880-500MG
101816-958EA 306.15 USD
101816-958
4-Amino-N-methyl-N-phenylbenzenesulfonamide
4-Amino-N-methyl-N-phenylbenzenesulfonamide

Hazard: Irritant

Formula: C₁₃H₁₄N₂O₂S
MDL Number: MFCD04576553
CAS Number: 63826-12-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR