4-[(4-Methyl-1-piperazinyl)sulfonyl]aniline
Supplier: Matrix Scientific
Synonyms:
4-((4-Methylpiperazin-1-yl)sulfonyl)aniline
, 4-((4-Methylpiperazin-1-yl)sulphonyl)aniline
023878-500MG
101816-956EA
81.54
USD
101816-956
4-[(4-Methyl-1-piperazinyl)sulfonyl]aniline
4-[(4-Methyl-1-piperazinyl)sulfonyl]aniline
Hazard: Irritant
Formula:
C₁₁H₁₇N₃O₂S MW: 255.34 g/mol |
MDL Number:
MFCD03050283 CAS Number: 21623-68-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...