4-Amino-N-(tert-butyl)benzenesulfonamide

Supplier: Matrix Scientific

Synonyms: N-tert-Butyl 4-Aminophenylsulfonamide

023876-500MG
101816-952EA 143.75 USD
101816-952
4-Amino-N-(tert-butyl)benzenesulfonamide
4-Amino-N-(tert-butyl)benzenesulfonamide

Hazard: Irritant

Formula: C₁₀H₁₆N₂O₂S
MW: 228.32 g/mol
MDL Number: MFCD03002128
CAS Number: 209917-48-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR