4-Amino-N-(4-ethoxyphenyl)benzenesulfonamide

Supplier: Matrix Scientific

023875-500MG
101816-950EA 216.92 USD
101816-950
4-Amino-N-(4-ethoxyphenyl)benzenesulfonamide
4-Amino-N-(4-ethoxyphenyl)benzenesulfonamide

Hazard: Irritant

Formula: C₁₄H₁₆N₂O₃S
MDL Number: MFCD02575821
CAS Number: 19837-75-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR