2-[1-(4-Ethoxybenzyl)-3-oxo-2-piperazinyl]acetic acid

Supplier: Matrix Scientific

023874-500MG
101816-948EA 228.85 USD
101816-948
2-[1-(4-Ethoxybenzyl)-3-oxo-2-piperazinyl]acetic acid
2-[1-(4-Ethoxybenzyl)-3-oxo-2-piperazinyl]acetic acid

Hazard: Irritant

Formula: C₁₅H₂₀N₂O₄
MDL Number: MFCD05666718
CAS Number: 1042696-40-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR