4-Amino-N-(2,4-dimethylphenyl)benzenesulfonamide

Supplier: Matrix Scientific

023871-500MG
101816-942EA 221.54 USD
101816-942
4-Amino-N-(2,4-dimethylphenyl)benzenesulfonamide
4-Amino-N-(2,4-dimethylphenyl)benzenesulfonamide

Hazard: Irritant

Formula: C₁₄H₁₆N₂O₂S
MDL Number: MFCD02575820
CAS Number: 327069-28-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR