Methyl-2-amino-5,6-dihydro-4H-cyclopenta[b]thiophene-3-carboxylate

Supplier: Matrix Scientific

023864-500MG
101816-928EA 11.5 USD
101816-928
Methyl-2-amino-5,6-dihydro-4H-cyclopenta[b]thiophene-3-carboxylate
Methyl-2-amino-5,6-dihydro-4H-cyclopenta[b]thiophene-3-carboxylate

Hazard: Irritant

Formula: C₉H₁₁NO₂S
MDL Number: MFCD00625612
CAS Number: 184174-80-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR