4-(2-Aminobenzoyl)-2-piperazinone

Supplier: Matrix Scientific

023863-500MG
101816-926EA 180 USD
101816-926
4-(2-Aminobenzoyl)-2-piperazinone
4-(2-Aminobenzoyl)-2-piperazinone

Hazard: Irritant

Formula: C₁₁H₁₃N₃O₂
MDL Number: MFCD05666702
CAS Number: 671794-74-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR