4-Amino-N-phenethylbenzenesulfonamide

Supplier: Matrix Scientific

Synonyms: C7280948

023862-500MG
101816-924EA 306.15 USD
101816-924
4-Amino-N-phenethylbenzenesulfonamide
4-Amino-N-phenethylbenzenesulfonamide

Hazard: Irritant

Formula: C₁₄H₁₆N₂O₂S
MDL Number: MFCD03716650
CAS Number: 587850-67-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR