4-Amino-N-phenylbenzenesulfonamide

023861-500MG
101816-922EA 203.79 USD
101816-922
4-Amino-N-phenylbenzenesulfonamide
4-Amino-N-phenylbenzenesulfonamide

Hazard: Irritant

Formula: C₁₂H₁₂N₂O₂S
MW: 248.31 g/mol
MDL Number: MFCD00233654
CAS Number: 127-77-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR