4-Amino-N-(3,4-dimethylphenyl)benzenesulfonamide

Supplier: Matrix Scientific

023860-500MG
101816-920EA 195.38 USD
101816-920
4-Amino-N-(3,4-dimethylphenyl)benzenesulfonamide
4-Amino-N-(3,4-dimethylphenyl)benzenesulfonamide

Hazard: Irritant

Formula: C₁₄H₁₆N₂O₂S
MDL Number: MFCD02188623
CAS Number: 20555-67-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR